missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-p-tolylmethane 99.0+%, TCI America™
Supplier: TCI America D131225ML
Specifications
| Di-p-tolylmethane | |
| White | |
| C15H16 | |
| 25 mL | |
| 4,4′C-Dimethyldiphenylmethane | |
| CC1=CC=C(C=C1)CC2=CC=C(C=C2)C | |
| 196.293 | |
| 196.29 | |
| Crystalline Lumps |
| 4957-14-6 | |
| 150°C | |
| MFCD00026018 | |
| 2811 | |
| HZAWPPRBCALFRN-UHFFFAOYSA-N | |
| 1-methyl-4-[(4-methylphenyl)methyl]benzene | |
| 21071 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 4957-14-6 | |
| 196.293 | |
| HZAWPPRBCALFRN-UHFFFAOYSA-N | |
| 21071 | |
| CC1=CC=C(C=C1)CC2=CC=C(C=C2)C |
| C15H16 | |
| MFCD00026018 | |
| 4,4′C-Dimethyldiphenylmethane | |
| 1-methyl-4-[(4-methylphenyl)methyl]benzene |
Safety and Handling
RTECSNumber : PA9100000
TSCA : No