accessibility menu, dialog, popup

UserName

CAS RN 4946-22-9

H3CNCH3SH

CAS RN 4946-22-9

IUPAC Name: 4-(dimethylamino)benzene-1-thiol
Synonyms: P-(dimethylamino)benzenethiol
Molecular Weight (g/mol): 153.24
Molecular Formula: C8H11NS
InChi Key: PQSBRHXGVPVYFJ-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(S)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results