CAS RN 492-98-8
IUPAC Name:
1H,1'H-2,2'-biimidazole
Synonyms:
2,2'-biimidazole
Molecular Weight (g/mol):
134.14
Molecular Formula:
C6H6N4
InChi Key:
AZUHIVLOSAPWDM-UHFFFAOYSA-N
SMILES:
N1C=CN=C1C1=NC=CN1
1
–
2
of
2
results
1
Promotions Available
2,2'-Biimidazole
2,2'-Biimidazole
CAS: 492-98-8 Molecular Formula: C6H6N4 Molecular Weight (g/mol): 134.14 MDL Number: MFCD00047014 InChI Key: AZUHIVLOSAPWDM-UHFFFAOYSA-N Synonym: 2,2 '-Bisimidazole; 2,2 '-Diimidazole IUPAC Name: 1H,1'H-2,2'-biimidazole SMILES: N1C=CN=C1C1=NC=CN1
2
2,2'-Biimidazole 98.0+%, TCI America™
CAS: 492-98-8 Molecular Formula: C6H6N4 Molecular Weight (g/mol): 134.14 MDL Number: MFCD00047014 InChI Key: AZUHIVLOSAPWDM-UHFFFAOYSA-N PubChem CID: 101463 IUPAC Name: 1H,1'H-2,2'-biimidazole SMILES: N1C=CN=C1C1=NC=CN1