missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-bromobutyrate, 98+%, Thermo Scientific™

Catalog No. p-2691752
Change view
Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC365830100 10g Glass bottle
AC365830500 50g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC365830100 Supplier Thermo Scientific Chemicals Supplier No. 365830100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 4897-84-1
Molecular Formula C5H9BrO2
Molecular Weight (g/mol) 181.03
MDL Number MFCD00041482
InChI Key QAWFLJGZSZIZHO-UHFFFAOYSA-N
Synonym methyl 4-bromobutyrate, 4-bromobutyric acid methyl ester, methyl-4-bromobutyrate, butanoic acid, 4-bromo-, methyl ester, methyl4-bromobutyrate, butyric acid, 4-bromo-, methyl ester, methyl gamma-bromobutyrate, acmc-1amn9, 4-brombutyric acid methyl ester, 4-bromobutyric acid methyl-ester
PubChem CID 107604
IUPAC Name methyl 4-bromobutanoate
SMILES COC(=O)CCCBr

Specifications

Density 1.4340g/mL
Boiling Point 186.0°C to 187.0°C
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.4610 to 1.4630 (20°C, 589nm)
Quantity 10g
Specific Gravity 1.434
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 181.03
Percent Purity 98+%
Chemical Name or Material Methyl 4-bromobutyrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Call a POISON CENTER or doctor/physicia
EINECSNumber 225-523-3

RUO – Research Use Only