missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-bromobutyrate, 97%

Catalog No. AAL1263618
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1263618 50 g
AAL1263609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1263618 Supplier Thermo Scientific Chemicals Supplier No. L1263618

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4897-84-1 | C5H9BrO2 | 181.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4897-84-1
Molecular Formula C5H9BrO2
Molecular Weight (g/mol) 181.03
MDL Number MFCD00041482
InChI Key QAWFLJGZSZIZHO-UHFFFAOYSA-N
Synonym methyl 4-bromobutyrate, 4-bromobutyric acid methyl ester, methyl-4-bromobutyrate, butanoic acid, 4-bromo-, methyl ester, methyl4-bromobutyrate, butyric acid, 4-bromo-, methyl ester, methyl gamma-bromobutyrate, acmc-1amn9, 4-brombutyric acid methyl ester, 4-bromobutyric acid methyl-ester
PubChem CID 107604
IUPAC Name methyl 4-bromobutanoate
SMILES COC(=O)CCCBr

Specifications

Boiling Point 186°C to 187°C
Refractive Index 1.462
Quantity 50 g
Beilstein 1745618
Formula Weight 181.04
Percent Purity 97%
Chemical Name or Material Methyl 4-bromobutyrate
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
EINECSNumber 225-523-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only