accessibility menu, dialog, popup

UserName

CAS RN 4897-50-1

IUPAC Name: 1,4'-bipiperidine
Synonyms: 4-piperidino-piperidine
Molecular Weight (g/mol): 168.28
Molecular Formula: C10H20N2
InChi Key: QDVBKXJMLILLLB-UHFFFAOYSA-N
SMILES: C1CCN(CC1)C1CCNCC1
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)