accessibility menu, dialog, popup

UserName

CAS RN 489-78-1

NH2OHSOHOO

CAS RN 489-78-1

IUPAC Name: 8-amino-4-hydroxynaphthalene-2-sulfonic acid
Synonyms: 8-amino-4-hydroxynaphthalene-2-sulfonic acid
Molecular Weight (g/mol): 239.25
Molecular Formula: C10H9NO4S
InChi Key: GGZZISOUXJHYOY-UHFFFAOYSA-N
SMILES: NC1=C2C=C(C=C(O)C2=CC=C1)S(O)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results