accessibility menu, dialog, popup

UserName

HOOOOH

CAS RN 486-35-1

IUPAC Name: 7,8-dihydroxy-2H-chromen-2-one
Synonyms: daphnetin daphnetol
Molecular Weight (g/mol): 178.14
Molecular Formula: C9H6O4
InChi Key: ATEFPOUAMCWAQS-UHFFFAOYSA-N
SMILES: OC1=CC=C2C=CC(=O)OC2=C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results