missing translation for 'onlineSavingsMsg'
Learn More

Daphnetin 90.0+%, TCI America™

Catalog No. D40015G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40015G 5 g
D40011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D40015G Supplier TCI America Supplier No. D40015G
Only null left

Chemical Identifiers

CAS 486-35-1
Molecular Formula C9H6O4
Molecular Weight (g/mol) 178.143
MDL Number MFCD00016977
InChI Key ATEFPOUAMCWAQS-UHFFFAOYSA-N
Synonym 7,8-Dihydroxycoumarin
PubChem CID 5280569
ChEBI CHEBI:17313
IUPAC Name 7,8-dihydroxychromen-2-one
SMILES C1=CC(=C(C2=C1C=CC(=O)O2)O)O

Specifications

Melting Point 256°C
Color White-Yellow
Quantity 5 g
Formula Weight 178.14
Percent Purity ≥90.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Daphnetin
RTECSNumber GN6382600
TSCA No