accessibility menu, dialog, popup

UserName

CAS RN 4857-06-1

IUPAC Name: 2-chloro-1H-1,3-benzodiazole
Synonyms: 2-chloro-1H-1,3-benzodiazole
Molecular Weight (g/mol): 152.58
Molecular Formula: C7H5ClN2
InChi Key: AYPSHJCKSDNETA-UHFFFAOYSA-N
SMILES: ClC1=NC2=CC=CC=C2N1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)