accessibility menu, dialog, popup

UserName

CAS RN 482-89-3

ONHHNO(Z)

CAS RN 482-89-3

IUPAC Name: (Z)-1H,1'H,3H,3'H-[2,2'-biindolylidene]-3,3'-dione
Synonyms: indigo blue
Molecular Weight (g/mol): 262.27
Molecular Formula: C16H10N2O2
InChi Key: COHYTHOBJLSHDF-YPKPFQOOSA-N
SMILES: O=C1\C(NC2=CC=CC=C12)=C1\NC2=CC=CC=C2C1=O
Quantity 
  • (1)
  • (2)
Color 
  • (2)
Percent Purity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)