missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Indigo, synthetic

Catalog No. AC212130250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC212130250 25 g Glass bottle
AC212131000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC212130250 Supplier Thermo Scientific Chemicals Supplier No. 212130250
Only null left

CAS: 482-89-3 | C16H10N2O2 | 262.268 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 482-89-3
Molecular Formula C16H10N2O2
Molecular Weight (g/mol) 262.268
MDL Number MFCD00005722
InChI Key COHYTHOBJLSHDF-BUHFOSPRSA-N
Synonym C.I. 73000, Vat Blue 1
PubChem CID 5318432
IUPAC Name (2E)-2-(3-oxo-1H-indol-2-ylidene)-1H-indol-3-one
SMILES C1=CC=C2C(=C1)C(=O)C(=C3C(=O)C4=CC=CC=C4N3)N2

Specifications

Beilstein 24, 417
Merck Index 15, 4981
Infrared Spectrum Authentic
Formula Weight 262.26
Packaging Glass bottle
UV Spectrometry Complies
Color Blue to Violet
Quantity 25 g
Physical Form Powder
Chemical Name or Material Indigo, synthetic
EINECSNumber 207-586-9
RTECSNumber DU2988400
TSCA TSCA

RUO – Research Use Only