missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phyproof™ Indigotin, ≥90% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 482-89-3 |
| Product Line | phyproof |
| Linear Formula | C16H10N2O2 |
| MDL Number | MFCD00005722 |
| Melting Point | >300°C |
| Synonym | Indigo; Indigo blue; Indigotin |
| Type | Reference Substance |
| Quantity | 10 mg |
| Formula Weight | 262.26 |
| Percent Purity | ≥90% (HPLC) |
| Show More |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG