missing translation for 'onlineSavingsMsg'
Learn More

Indigo (synthetic) 97.0+%, TCI America™

Catalog No. I021225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I021225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I021225G Supplier TCI America Supplier No. I021225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 482-89-3
Molecular Formula C16H10N2O2
Molecular Weight (g/mol) 262.268
MDL Number MFCD00005722
InChI Key COHYTHOBJLSHDF-BUHFOSPRSA-N
Synonym indigo, indigotin, cystoceva, diindogen, vulcafix blue r, pigment blue 66, vat blue 1, indigo ciba, indigo synthetic, synthetic indigo
PubChem CID 5318432
IUPAC Name (2E)-2-(3-oxo-1H-indol-2-ylidene)-1H-indol-3-one
SMILES C1=CC=C2C(=C1)C(=O)C(=C3C(=O)C4=CC=CC=C4N3)N2

Specifications

Color Blue-Purple
Quantity 25 g
Formula Weight 262.27
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Indigo (synthetic)
EINECSNumber (5)-2223
RTECSNumber DU2988400
TSCA Yes