accessibility menu, dialog, popup

UserName

CAS RN 4779-94-6

HClNH2OHHO

CAS RN 4779-94-6

IUPAC Name: hydrogen 3-(2-amino-1-hydroxyethyl)phenol chloride
Synonyms: norfenefrine hydrochloride
Molecular Weight (g/mol): 189.64
Molecular Formula: C8H12ClNO2
InChi Key: OWMFSWZUAWKDRR-UHFFFAOYNA-N
SMILES: [H+].[Cl-].NCC(O)C1=CC=CC(O)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results