missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Norphenylephrine Hydrochloride 98.0+%, TCI America™
$127.07 - $336.90
Chemical Identifiers
| CAS | 4779-94-6 |
|---|---|
| Molecular Formula | C8H12ClNO2 |
| Molecular Weight (g/mol) | 189.639 |
| MDL Number | MFCD00012879 |
| InChI Key | OWMFSWZUAWKDRR-UHFFFAOYSA-N |
| Synonym | 3-(2-Amino-1-hydroxyethyl)phenol Hydrochloride |
| PubChem CID | 3016408 |
| IUPAC Name | 3-(2-amino-1-hydroxyethyl)phenol;hydrochloride |
| SMILES | C1=CC(=CC(=C1)O)C(CN)O.Cl |
Chemical Identifiers
| 4779-94-6 | |
| 189.639 | |
| OWMFSWZUAWKDRR-UHFFFAOYSA-N | |
| 3016408 | |
| C1=CC(=CC(=C1)O)C(CN)O.Cl |
| C8H12ClNO2 | |
| MFCD00012879 | |
| 3-(2-Amino-1-hydroxyethyl)phenol Hydrochloride | |
| 3-(2-amino-1-hydroxyethyl)phenol;hydrochloride |
Specifications
| 4779-94-6 | |
| White | |
| MFCD00012879 | |
| 3-(2-Amino-1-hydroxyethyl)phenol Hydrochloride | |
| C1=CC(=CC(=C1)O)C(CN)O.Cl | |
| 189.639 | |
| 189.64 | |
| Crystalline Powder |
| 157°C | |
| C8H12ClNO2 | |
| 5 g | |
| OWMFSWZUAWKDRR-UHFFFAOYSA-N | |
| 3-(2-amino-1-hydroxyethyl)phenol;hydrochloride | |
| 3016408 | |
| ≥98.0% (T) | |
| DL-Norphenylephrine Hydrochloride |
Safety and Handling
RTECSNumber : DN7360000
TSCA : No