missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Norphenylephrine Hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| Packaging | Ampule |
| CAS | 4779-94-6 |
| Linear Formula | HOC6H4 CH(CH2 NH2)OH · HCl |
| MDL Number | MFCD00012879 |
| Melting Point | 162°C to 164°C |
| Synonym | Norphenylephrine hydrochloride; alpha-Aminomethyl-3-hydroxybenzyl alcohol hydrochloride; 3,alpha-Dihydroxyphenethylamine hydrochloride; Norfenefrine hydrochloride |
| Type | Certified Reference Material |
| Quantity | 200 mg |
| Formula Weight | 189.64 |
| Grade | Certified Reference Material |