accessibility menu, dialog, popup

UserName

CAS RN 471930-01-5

IUPAC Name: 3-[(piperidin-1-yl)methyl]benzaldehyde
Synonyms: 3-(piperidin-1-ylmethyl)benzaldehyde
Molecular Weight (g/mol): 203.29
Molecular Formula: C13H17NO
InChi Key: UJDTVJKIOWAUNQ-UHFFFAOYSA-N
SMILES: O=CC1=CC(CN2CCCCC2)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)