accessibility menu, dialog, popup

UserName

H3COOCH2

CAS RN 4704-31-8

IUPAC Name: ethenyl decanoate
Synonyms: ethenyl decanoate
Molecular Weight (g/mol): 198.31
Molecular Formula: C12H22O2
InChi Key: CMDXMIHZUJPRHG-UHFFFAOYSA-N
SMILES: CCCCCCCCCC(=O)OC=C
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)