missing translation for 'onlineSavingsMsg'
Learn More

Vinyl decanoate, 95%, stab.

Catalog No. AAH2908114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2908114 25 g
AAH2908106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2908114 Supplier Thermo Scientific Chemicals Supplier No. H2908114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4704-31-8 | C12H22O2 | 198.306 g/mol

It is employed as a intermediates for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediates for pharmaceutical.

Solubility
Soluble in methanol. Not miscible or difficult to mix with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 4704-31-8
Molecular Formula C12H22O2
Molecular Weight (g/mol) 198.306
MDL Number MFCD00027349
InChI Key CMDXMIHZUJPRHG-UHFFFAOYSA-N
Synonym vinyl decanoate, decanoic acid, vinyl ester, vinyl n-decanoate, unii-ff00i399li, decanoic acid, ethenyl ester, n-capric acid vinyl ester, decanoic acid vinyl, acmc-1aqz4, decanoic acid vinyl ester, capric acid vinyl ester = vinyl decenoate
PubChem CID 62140
IUPAC Name ethenyl decanoate
SMILES CCCCCCCCCC(=O)OC=C

Specifications

Density 0.886
Boiling Point 119°C to 120°C (10 mmHg)
Flash Point 104°C (219°F)
Refractive Index 1.435
Linear Formula CH3(CH2)8CO2CH=CH2
Quantity 25 g
Solubility Information Soluble in methanol. Not miscible or difficult to mix with water.
Formula Weight 198.3
Percent Purity 95%
Chemical Name or Material Vinyl decanoate, stabilized
EINECSNumber 225-185-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only