accessibility menu, dialog, popup

UserName

CAS RN 470-82-6

IUPAC Name: 1,3,3-trimethyl-2-oxabicyclo[2.2.2]octane
Synonyms: eucalyptol eucalyptole limonene oxide eucalyptol preparation 1,8-oxido-p-menthane cajeputol p-cineole eucalyptol (usan) CNL eucalyptus citriodora leaf extract
Molecular Weight (g/mol): 154.25
Molecular Formula: C10H18O
InChi Key: WEEGYLXZBRQIMU-UHFFFAOYSA-N
SMILES: CC12CCC(CC1)C(C)(C)O2
Boiling Point 
  • (4)
  • (5)
Percent Purity 
  • (3)
  • (1)
  • (5)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (5)
  • (4)
  • (5)
  • (3)
  • (1)
  • (5)
  • (9)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)