missing translation for 'onlineSavingsMsg'
Learn More

1,8-Cineole, 99%

Catalog No. AC110345000
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC110345000 500 g Glass bottle
AC110340050 5 g Glass bottle
AC110341000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC110345000 Supplier Thermo Scientific Chemicals Supplier No. 110345000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 470-82-6 | C10H18O | 154.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 470-82-6
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00167977
InChI Key WEEGYLXZBRQIMU-UHFFFAOYSA-N
Synonym eucalyptol, cineole, 1,8-cineole, 1,8-cineol, cajeputol, 1,8-epoxy-p-menthane, eucalyptole, zineol, eucapur, terpan
PubChem CID 2758
IUPAC Name 2,2,4-trimethyl-3-oxabicyclo[2.2.2]octane
SMILES CC12CCC(CC1)C(C)(C)O2

Specifications

Melting Point 1.5°C
Color Colorless to Yellow
Density 0.9225g/mL
Boiling Point 176°C to 177°C
Flash Point 49°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4550 to 1.46
Quantity 500 g
Beilstein 17,II,32
Specific Gravity 0.9225
Merck Index 15, 3938
Specific Rotation -0.3° to +0.3° (20°C, 589nm) (neat)
Solubility Information (in ethanol (70% v/v)) 1:2
Formula Weight 154.25
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1,8-Cineole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
EINECSNumber 207-431-5

RUO – Research Use Only