missing translation for 'onlineSavingsMsg'
Learn More

Eucalyptol (cineole) Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111018859
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018859 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018859 Supplier Merck Emd Millipore Supplier No. PHR1828500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 470-82-6
Density 0.921 g/mL (at 25°C)
Flash Point 49°C - closed cup
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C10H18 O
MDL Number MFCD00167977
Melting Point 1°C to 2°C
Boiling Point 176°C to 177°C
Synonym Eucalyptol; 1,3,3-Trimethyl-2-oxabicyclo[2.2.2 ]octane; 1,8-Cineole; 1,8-Epoxy-p-menthane
Type Certified Reference Material
Quantity 500 mg
Formula Weight 154.25
Grade Certified Reference Material
Show More Show Less