CAS RN 4670-05-7
IUPAC Nor:
—
Synonymes:
—
Poids moléculaire (g/mol):
—
Formule moléculaire:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
résultats
1
Theaflavin, ≥90% (HPLC), MilliporeSigma™ Supelco™
Theaflavin is a polyphenol, naturally available in tea, which can mainly play a role in the inhibition of xanthine oxidase enzyme to yield uric acid. It can also find applications in reducing oxidative stress and can also act like scavengers of superoxide.
2
phyproof™ Theaflavin, ≥95% (HPLC), MilliporeSigma™ Supelco™
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG