missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Theaflavin, ≥90% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 4670-05-7 |
| Physical Form | Neat |
| Linear Formula | C29H24O12 |
| MDL Number | MFCD03427500 |
| Synonym | 3,4,5-Trihydroxy-1,8-bis[(2 R,3 R)-3,5,7-trihydroxy-2-chromanyl]-6-benzo[7 ]annulenone; 3,4,6-Trihydroxy-1,8-bis(3 alpha,5,7-trihydroxy-2 alpha-chromanyl)-5 H-benzocyclohepten-5-one |
| Type | Analytical Standard |
| Quantity | 1 mg |
| Formula Weight | 564.49 |
| Percent Purity | ≥90% (HPLC) |
| Grade | Analytical Standard |