CAS RN 465514-09-4
IUPAC Name:
2-(morpholin-4-yl)pyridine-3-carbaldehyde
Synonyms:
2-(morpholin-4-yl)pyridine-3-carbaldehyde
Molecular Weight (g/mol):
192.22
Molecular Formula:
C10H12N2O2
InChi Key:
CCPVZIBHRFCNTR-UHFFFAOYSA-N
SMILES:
O=CC1=C(N=CC=C1)N1CCOCC1
1
–
1
of
1
results
1
2-Morpholin-4-yl-pyridine-3-carbaldehyde, 95%, Thermo Scientific™
CAS: 465514-09-4 Molecular Formula: C10H12N2O2 Molecular Weight (g/mol): 192.22 MDL Number: MFCD03086195 InChI Key: CCPVZIBHRFCNTR-UHFFFAOYSA-N Synonym: 2-morpholinonicotinaldehyde,2-morpholin-4-yl-pyridine-3-carbaldehyde,2-morpholin-4-ylpyridine-3-carboxaldehyde,2-morpholin-4-yl pyridine-3-carbaldehyde,2-morpholinopyridine-3-carboxaldehyde,2-morpholin-4-ylnicotinaldehyde,2-morpholin-4-yl nicotinaldehyde,2-morpholinopyridine-3-carbaldehyde,3-pyridinecarboxaldehyde,2-4-morpholinyl,3-pyridinecarboxaldehyde, 2-4-morpholinyl PubChem CID: 2776570 IUPAC Name: 2-morpholin-4-ylpyridine-3-carbaldehyde SMILES: O=CC1=C(N=CC=C1)N1CCOCC1