missing translation for 'onlineSavingsMsg'
Learn More

2-Morpholin-4-yl-pyridine-3-carbaldehyde, 95%, Thermo Scientific™

Numéro de catalogue. CC24604CB
Modifier l'affichage
Click to view available options
Quantité:
250mg
Emballage:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
CC24604CB 250mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. CC24604CB Fournisseur Thermo Scientific Chemicals Code fournisseur CC24604CB

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 465514-09-4
Formule moléculaire C10H12N2O2
Poids moléculaire (g/mol) 192.22
Numéro MDL MFCD03086195
Clé InChI CCPVZIBHRFCNTR-UHFFFAOYSA-N
Synonyme 2-morpholinonicotinaldehyde, 2-morpholin-4-yl-pyridine-3-carbaldehyde, 2-morpholin-4-ylpyridine-3-carboxaldehyde, 2-morpholin-4-yl pyridine-3-carbaldehyde, 2-morpholinopyridine-3-carboxaldehyde, 2-morpholin-4-ylnicotinaldehyde, 2-morpholin-4-yl nicotinaldehyde, 2-morpholinopyridine-3-carbaldehyde, 3-pyridinecarboxaldehyde,2-4-morpholinyl, 3-pyridinecarboxaldehyde, 2-4-morpholinyl
PubChem CID 2776570
Nom de l’IUPAC 2-morpholin-4-ylpyridine-3-carbaldehyde
SOURIRES O=CC1=C(N=CC=C1)N1CCOCC1

Spécifications

Nom ou substance chimique 2-Morpholinonicotinaldehyde
CAS % minimum 94.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 95%
Emballage Amber Glass Bottle
Quantité 250mg
Poids de formule 192.22
Pourcentage de pureté 95%
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.