accessibility menu, dialog, popup

UserName

CAS RN 463-04-7

IUPAC Name: pentyl nitrite
Synonyms: amyl nitrite 1-nitropentane
Molecular Weight (g/mol): 117.15
Molecular Formula: C5H11NO2
InChi Key: CSDTZUBPSYWZDX-UHFFFAOYSA-N
SMILES: CCCCCON=O
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results