missing translation for 'onlineSavingsMsg'
Learn More

Amyl Nitrite 95.0+%, TCI America™

Catalog No. A045525ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A045525ML 25 mL
A0455100ML 100 mL
A0455500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A045525ML Supplier TCI America Supplier No. A045525ML

Chemical Identifiers

CAS 463-04-7
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.148
MDL Number MFCD00036132
InChI Key CSDTZUBPSYWZDX-UHFFFAOYSA-N
Synonym Pentyl Nitrite
PubChem CID 10026
ChEBI CHEBI:55344
IUPAC Name pentyl nitrite
SMILES CCCCCON=O

Specifications

Color Yellow
Boiling Point 104°C
Quantity 25 mL
UN Number 1113
Formula Weight 117.15
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Amyl Nitrite
RTECSNumber RA1140000
TSCA Yes
Recommended Storage Refrigerator