accessibility menu, dialog, popup

UserName

H2NNH2

CAS RN 462-94-2

IUPAC Name: pentane-1,5-diamine
Synonyms: pentane-1,5-diamine cadaverine (substance) animal coniine BioDex 1 DAPE
Molecular Weight (g/mol): 102.18
Molecular Formula: C5H14N2
InChi Key: VHRGRCVQAFMJIZ-UHFFFAOYSA-N
SMILES: NCCCCCN
Percent Purity 
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)