missing translation for 'onlineSavingsMsg'
Learn More

Cadaverine, Analytical Standard, ≥96.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019431
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019431 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019431 Supplier Merck Emd Millipore Supplier No. 520631ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 462-94-2
Density 0.873 g/mL (at 25°C)
Physical Form Neat
Linear Formula NH2(CH2)5 NH2
MDL Number MFCD00008239
Melting Point 14°C to 16°C
Boiling Point 178°C to 180°C
Synonym 1,5-Diaminopentane; 1,5-Pentanediamine; Pentamethylenediamine
Quantity 1 mL
Formula Weight 102.18
Percent Purity ≥96.5% (GC)
Grade Analytical Standard
Show More Show Less