accessibility menu, dialog, popup

UserName

CAS RN 45676-04-8

CH3H3CCH3NN

CAS RN 45676-04-8

IUPAC Name: 1-tert-butyl-1H-imidazole
Synonyms: 1-tert-butylimidazole
Molecular Weight (g/mol): 124.19
Molecular Formula: C7H12N2
InChi Key: AMQKPABOPFXDQM-UHFFFAOYSA-N
SMILES: CC(C)(C)N1C=CN=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results