accessibility menu, dialog, popup

UserName

CAS RN 45644-21-1

IUPAC Name: 6-chloropyridin-2-amine
Synonyms: 6-chloropyridin-2-amine
Molecular Weight (g/mol): 128.56
Molecular Formula: C5H5ClN2
InChi Key: OBYJTLDIQBWBHM-UHFFFAOYSA-N
SMILES: NC1=CC=CC(Cl)=N1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (1)
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (3)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (4)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)