missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-chloropyridine 98.0+%, TCI America™

Catalog No. A22635G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A22635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A22635G Supplier TCI America Supplier No. A22635G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Synthetic building block with a minimum purity of 98.0% by GC and nonaqueous titration.

Chemical Identifiers

CAS 45644-21-1
Molecular Formula C5H5ClN2
Molecular Weight (g/mol) 128.559
MDL Number MFCD00234068
InChI Key OBYJTLDIQBWBHM-UHFFFAOYSA-N
Synonym 2-amino-6-chloropyridine, 2-chloro-6-aminopyridine, 2-pyridinamine, 6-chloro, 6-chloro-2-pyridinamine, 6-chloro-pyridin-2-ylamine, pyridine, 2-amino-6-chloro, 6-chloropyridin-2-ylamine, 6-chloro-2-pyridylamine, 6-amino-2-chloropyridine, 2-amino-6-chloropyridine 6-chloropyridin-2-ylamine
PubChem CID 206246
IUPAC Name 6-chloropyridin-2-amine
SMILES C1=CC(=NC(=C1)Cl)N

Specifications

Melting Point 72°C
Quantity 5 g
Formula Weight 128.56
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-6-chloropyridine
EINECSNumber (5)-5826
RTECSNumber US1813350
TSCA No