accessibility menu, dialog, popup

UserName

CAS RN 4562-27-0

IUPAC Name: 3,4-dihydropyrimidin-4-one
Synonyms: pyrimidone
Molecular Weight (g/mol): 96.09
Molecular Formula: C4H4N2O
InChi Key: DNCYBUMDUBHIJZ-UHFFFAOYSA-N
SMILES: O=C1NC=NC=C1
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)