accessibility menu, dialog, popup

UserName

CAS RN 456-64-4

IUPAC Name: 1,1,1-trifluoro-N-phenylmethanesulfonamide
Synonyms: CF3-methanesulfonanilide
Molecular Weight (g/mol): 225.19
Molecular Formula: C7H6F3NO2S
InChi Key: OXDSKEQSEGDAFN-UHFFFAOYSA-N
SMILES: FC(F)(F)S(=O)(=O)NC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results