missing translation for 'onlineSavingsMsg'
Learn More

Trifluoromethanesulfonanilide 98.0+%, TCI America™

Catalog No. T29855G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T29855G 5 g
T29851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T29855G Supplier TCI America Supplier No. T29855G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 456-64-4
Molecular Formula C7H6F3NO2S
Molecular Weight (g/mol) 225.185
MDL Number MFCD00521820
InChI Key OXDSKEQSEGDAFN-UHFFFAOYSA-N
Synonym N-Phenyl(trifluoromethane)sulfonamide, 1,1,1-Trifluoro-N-phenylmethanesulfonamide
PubChem CID 68012
IUPAC Name 1,1,1-trifluoro-N-phenylmethanesulfonamide
SMILES C1=CC=C(C=C1)NS(=O)(=O)C(F)(F)F

Specifications

Melting Point 67°C
Color White
Quantity 5 g
UN Number 2811
Formula Weight 225.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Trifluoromethanesulfonanilide
RTECSNumber PB0482000
TSCA No