accessibility menu, dialog, popup

UserName

ONONNFBFFF

CAS RN 456-27-9

IUPAC Name: 4-nitrobenzene-1-diazonium; tetrafluoroboranuide
Synonyms: 4-nitrobenzenediazonium tetrafluoroborate
Molecular Weight (g/mol): 236.92
Molecular Formula: C6H4BF4N3O2
InChi Key: AFULQCYCQAHYIP-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.[O-][N+](=O)C1=CC=C(C=C1)[N+]#N
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)