missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzenediazonium Tetrafluoroborate 98.0+%, TCI America™

Catalog No. N013725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N013725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N013725G Supplier TCI America Supplier No. N013725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 456-27-9
Molecular Formula C6H4BF4N3O2
Molecular Weight (g/mol) 236.92
MDL Number MFCD00012005
InChI Key AFULQCYCQAHYIP-UHFFFAOYSA-N
Synonym 4-nitrobenzenediazonium tetrafluoroborate, ccris 3641, unii-2uxo615r4o, p-nitrobenzenediazonium tetrafluoroborate, benzenediazonium, 4-nitro-, tetrafluoroborate 1-, p-nitrophenyldiazonium fluoborate, p-nitrobenzenediazonium fluoborate, 4-nitrobenzenediazonium fluoborate, p-nitrobenzenediazonium fluoroborate, 4-nitrobenzenediazonium fluoroborate
PubChem CID 68006
IUPAC Name 4-nitrobenzene-1-diazonium; tetrafluoroboranuide
SMILES F[B-](F)(F)F.[O-][N+](=O)C1=CC=C(C=C1)[N+]#N

Specifications

Color White-Yellow
Quantity 25 g
UN Number 1759
Formula Weight 236.92
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrobenzenediazonium Tetrafluoroborate
RTECSNumber CZ1723000
TSCA Yes
Recommended Storage Freezer