accessibility menu, dialog, popup

UserName

NFFFFFF

CAS RN 455-00-5

IUPAC Name: 2,6-bis(trifluoromethyl)pyridine
Synonyms: 2,6-bis(trifluoromethyl)pyridine
Molecular Weight (g/mol): 215.1
Molecular Formula: C7H3F6N
InChi Key: YPDVFTXBESQIPJ-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC(=N1)C(F)(F)F
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)