missing translation for 'onlineSavingsMsg'
Learn More

2,6-Bis(trifluoromethyl)pyridine 98.0+%, TCI America™

Catalog No. B23951G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B23951G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B23951G Supplier TCI America Supplier No. B23951G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 455-00-5
Molecular Formula C7H3F6N
Molecular Weight (g/mol) 215.098
MDL Number MFCD00236675
InChI Key YPDVFTXBESQIPJ-UHFFFAOYSA-N
Synonym 2,6-bis trifluoromethyl pyridine, 2,6-bis trifluoromethyl-pyridine, pubchem7696, acmc-209k2t, pyridine,2,6-bis trifluoromethyl, pyridine, 2,6-bis trifluoromethyl
PubChem CID 589864
IUPAC Name 2,6-bis(trifluoromethyl)pyridine
SMILES C1=CC(=NC(=C1)C(F)(F)F)C(F)(F)F

Specifications

Melting Point 57°C
Color White-Yellow
Boiling Point 154°C
Quantity 1 g
Formula Weight 215.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Bis(trifluoromethyl)pyridine