accessibility menu, dialog, popup

UserName

CAS RN 4548-53-2

NaNaCH3H3CHNNOSOOOSOOO(E)

CAS RN 4548-53-2

IUPAC Name: disodium (3E)-3-[2-(2,4-dimethyl-5-sulfonatophenyl)hydrazin-1-ylidene]-4-oxo-3,4-dihydronaphthalene-1-sulfonate
Synonyms: scarlet GN
Molecular Weight (g/mol): 480.42
Molecular Formula: C18H14N2Na2O7S2
InChi Key: GRRDBTDTMCTWQZ-ZAWKPCBGSA-L
SMILES: [Na+].[Na+].CC1=CC(C)=C(C=C1N\N=C1/C=C(C2=CC=CC=C2C1=O)S([O-])(=O)=O)S([O-])(=O)=O
Grade 
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Color 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results