missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D and C Red No. 4, 87%, Spectrum™ Chemical
Description
Spectrum™ Chemical D and C Red No. 4 is a certified color widely used in cleaning compounds and personal care products. Some of the more widely used applications for certified dyes include shampoos, soaps, make-up, and cleaning products. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Specifications
Specifications
| CAS Max % | 100% |
| Percent Purity | 87% |
| Grade | Reagent |
| Packaging | Amber Glass Bottle |
| Color | Red/Brown to Dark Red |
| Quantity | 500 g |
| Chemical Name or Material | D and C Red No. 4 |