missing translation for 'onlineSavingsMsg'
Learn More

Ponceau SX, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014220
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014220 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014220 Supplier Merck Emd Millipore Supplier No. 9239525MG
Only null left

Ponceau SX is classified as a synthetic food color, widely used for the maintenance and improvement of color appearance in foods.

Specifications

CAS 4548-53-2
Physical Form Neat
Linear Formula C18H14N2 Na2O7S2
MDL Number MFCD00046402
Synonym 3-(2,4-Dimethyl-5-sulfophenylazo)-4-hydroxynaphthalene-1-sulfonic acid disodium salt; Food Red 1
Type Analytical Standard
Quantity 25 mg
Formula Weight 480.42
Percent Purity ≥98% (HPLC)
Grade Analytical Standard