accessibility menu, dialog, popup

UserName

POHOOHPOHOOH

CAS RN 4546-06-9

IUPAC Name: {[4-(phosphonomethyl)phenyl]methyl}phosphonic acid
Synonyms: [4-(phosphonomethyl)phenyl]methylphosphonic acid
Molecular Weight (g/mol): 266.13
Molecular Formula: C8H12O6P2
InChi Key: ZURHBENZJDSCRG-UHFFFAOYSA-N
SMILES: OP(O)(=O)CC1=CC=C(CP(O)(O)=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results