accessibility menu, dialog, popup

UserName

CAS RN 45438-80-0

IUPAC Name: bromozincylium; thiophen-2-ide
Synonyms: bromozincylium; thiophen-2-ide
Molecular Weight (g/mol): 228.41
Molecular Formula: C4H3BrSZn
InChi Key: KRDONBTWAGIZME-UHFFFAOYSA-M
SMILES: [Zn+]Br.S1C=CC=[C-]1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results