missing translation for 'onlineSavingsMsg'
Learn More

2-Thienylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

Catalog No. AAH58397AD
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH58397AD 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH58397AD Supplier Thermo Scientific Chemicals Supplier No. H58397AD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 45438-80-0 | C4H3BrSZn | 228.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Thienylzinc bromide is used as medical intermediates.

Solubility
Reacts with water.

Notes
Air Sensitive. Store at 4°C. Incompatible with oxidizing agents, acid chlorides. Store under dry inert

Chemical Identifiers

CAS 45438-80-0
Molecular Formula C4H3BrSZn
Molecular Weight (g/mol) 228.41
MDL Number MFCD01317002
InChI Key KRDONBTWAGIZME-UHFFFAOYSA-M
Synonym zinc 2h-thiophen-2-ide bromide, zinc 2+ ion thiophen-2-ide bromide
PubChem CID 4428324
IUPAC Name bromozinc(1+);2H-thiophen-2-ide
SMILES [Zn+]Br.S1C=CC=[C-]1

Specifications

Density 0.973
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive
Solubility Information Reacts with water.
Formula Weight 228.41
Chemical Name or Material 2-Thienylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles
Hazard Category H225-H302-H314-H335-H336-H351
Hazard Statement GHS H Statement
H225-H261-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only