accessibility menu, dialog, popup

UserName

CAS RN 4472-44-0

IUPAC Name: 2-chloro-4,6-dimethylpyrimidine
Synonyms: 2-chloro-4,6-dimethylpyrimidine
Molecular Weight (g/mol): 142.59
Molecular Formula: C6H7ClN2
InChi Key: RZVPFDOTMFYQHR-UHFFFAOYSA-N
SMILES: CC1=CC(C)=NC(Cl)=N1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)