missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4,6-dimethylpyrimidine 98.0+%, TCI America™

Catalog No. C21995G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C21995G 5 g
C219925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C21995G Supplier TCI America Supplier No. C21995G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4472-44-0
Molecular Formula C6H7ClN2
Molecular Weight (g/mol) 142.586
MDL Number MFCD00023199
InChI Key RZVPFDOTMFYQHR-UHFFFAOYSA-N
Synonym pyrimidine, 2-chloro-4,6-dimethyl, 2-chloro-4,6-dimethyl-pyrimidine, pubchem6904, acmc-1adw7, 5-23-05-00401 beilstein handbook reference, ksc495i5r, 2-chlor-4,6-dimethylpyrimidine, 4,6-dimethyl-2-chloropyrimidine, 2-chloro-4,6-dimethyl-1,3-diazine, 2-chloro-4,6-dimethylpyrimidine
PubChem CID 20550
IUPAC Name 2-chloro-4,6-dimethylpyrimidine
SMILES CC1=CC(=NC(=N1)Cl)C

Specifications

Melting Point 37°C
Boiling Point 212°C
Quantity 5 g
Formula Weight 142.59
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4,6-dimethylpyrimidine
RTECSNumber UV8000000
TSCA No
Recommended Storage Refrigerator