CAS RN 446866-87-1
IUPAC Name:
3-(morpholin-4-yl)benzaldehyde
Synonyms:
3-(morpholin-4-yl)benzaldehyde
Molecular Weight (g/mol):
191.23
Molecular Formula:
C11H13NO2
InChi Key:
LQORKFSMUOSSQM-UHFFFAOYSA-N
SMILES:
O=CC1=CC(=CC=C1)N1CCOCC1
1
–
3
of
3
results
3
3-morpholinobenzaldehyde, 97%, Thermo Scientific™
CAS: 446866-87-1 Molecular Formula: C11H13NO2 Molecular Weight (g/mol): 191.23 InChI Key: LQORKFSMUOSSQM-UHFFFAOYSA-N Synonym: 3-morpholinobenzaldehyde,3-morpholin-4-yl benzaldehyde,4-3-formylphenyl morpholine,3-4-morpholinyl benzaldehyde,3-morpholin-4-yl-benzaldehyde,pubchem13334,3-morpholin-4-yl benzaldehyd,benzaldehyde,3-4-morpholinyl PubChem CID: 7164584 IUPAC Name: 3-morpholin-4-ylbenzaldehyde SMILES: C1COCCN1C2=CC=CC(=C2)C=O